Learn More
Alkali Scientific Ninhydrin Reagent, 100 G
SDP
Supplier:  Alkali Scientific NIN001CP
Encompass
Ninhydrin Reagent, 100g is a chemical reagent primarily used for the detection of amino acids, peptides, and proteins. When applied, ninhydrin reacts with free amines, turning a purple color, which is commonly used for amino acid quantification in chromatography and electrophoresis. This reagent is widely utilized in laboratory settings for detecting and analyzing amino acids in biological samples, such as urine, blood, and tissue extracts. The 100g pack size is suitable for high-throughput laboratories and research facilities conducting multiple experiments or large-scale studies in proteomics and amino acid analysis. It plays an essential role in protein research and metabolic studies.
Catalog No. 50-006-61466