Learn More
Medchemexpress LLC Nintedanib-13C,d3 | 656247-17-5 | 99.8% | C3013CH30D3N5O4 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY50904S11MG
Encompass_Preferred
Nintedanib-13C,d3 is a 13C- and deuterium labeled version of Nintedanib, a potent triple angiokinase inhibitor targeting VEGFR1/2/3, FGFR1/2/3, and PDGFRα/β. It is intended for research use only.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Potent triple angiokinase inhibitor.
Catalog No. 50-003-58415