Learn More
Medchemexpress LLC HY-50904 500mg , Nintedanib CAS:656247-17-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY50904500MG
Encompass_Preferred
HY-50904 500mg Nintedanib CAS: 656247-17-5 Nintedanib (BIBF 1120) is a potent triple angiokinase inhibitor for VEGFR1/2/3, FGFR1/2/3 and PDGFRα/β with IC50s of 34 nM/13 nM/13 nM, 69 nM/37 nM/108 nM and 59 nM/65 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9005