Learn More
Enzo Life Sciences Nintedanib. ethanesulfonate salt (1g). CAS: 656247-18-6
SDP
Supplier:  Enzo Life Sciences ENZCHM2291000
Encompass_Preferred
Nintedanib is a potent inhibitor of multiple receptor tyrosine kinases (RTKs) and non-receptor tyrosine kinases (nRTKs). Purity: ≥99% (HPLC). Solubility: Soluble in DMSO (25 mg/mL). Long Term Storage: -20°C.
Catalog No. 50-201-3626