Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nintedanib impurity 5 | 676326-36-6 | C₁₂H₁₁NO₄ | 50 MG
SDP

Supplier:  Medchemexpress LLC HY7569750MG

Encompass_Preferred

Nintedanib impurity 5 is an impurity of Nintedanib.

  • CAS Number: 676326-36-6
  • Molecular Formula: C₁₂H₁₁NO₄
  • Molecular Weight: 233.22
  • Storage: Store at room temperature for 3 years; in solvent -80°C for 2 years; -20°C for 1 year.

Catalog No. 50-004-33613


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.