Learn More
Medchemexpress LLC Nintedanib impurity 5 | 676326-36-6 | C12H11NO4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY75697100MG
Encompass_Preferred
Nintedanib impurity 5 is an impurity of Nintedanib (HY-50904), a target drug intermediate. This compound is suitable for various laboratory applications.
  • Stable at room temperature for 3 years
  • Stable in solvent at -80°C for 2 years
  • Stable in solvent at -20°C for 1 year
Catalog No. 50-004-34338