Learn More
Medchemexpress LLC Nintedanib impurity 5 | 676326-36-6 | C12H11NO4 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY75697
Encompass_Preferred
Nintedanib impurity 5 is an impurity of Nintedanib. This product is intended for research use only and has not been fully validated for medical applications.
  • Store at room temperature for 3 years.
  • When in solvent, store at -80°C for 2 years.
  • When in solvent, store at -20°C for 1 year.
Catalog No. 50-003-59262