Learn More
Medchemexpress LLC Nipecotic acid | 498-95-3 | 97.0% | C6H11NO2 | 5 G
SDP
Supplier:  Medchemexpress LLC HY693595G
Encompass_Preferred
Nipecotic acid is a potent inhibitor of neuronal and glial-aminobutyric acid (GABA) uptake in vitro. It can also directly activate GABAA-like chloride channels, with an EC50 of approximately 300 μM. This compound does not readily cross the blood-brain barrier and is intended for research use only.
  • Potent inhibitor of neuronal and glial-aminobutyric acid (GABA) uptake in vitro
  • Directly activates GABAA-like chloride channels
  • Does not readily cross the blood-brain barrier
  • Intended for research use only
Catalog No. 50-004-33792