Learn More
Medchemexpress LLC Nipecotic acid | 498-95-3 | ≥98.0% | C6H11NO2 | 1 G
SDP
Supplier:  Medchemexpress LLC HY693591G
Encompass_Preferred
Nipecotic acid ((±)-β-Homoproline) is a potent inhibitor of neuronal and glial-aminobutyric acid (GABA) uptake in vitro. It also directly activates GABAA-like chloride channels.
  • Potent inhibitor of neuronal and glial-aminobutyric acid (GABA) uptake in vitro
  • Directly activates GABAA-like chloride channels
  • Has an EC50 of approximately 300 μM
  • Does not readily cross the blood-brain barrier
Catalog No. 50-004-33578