Learn More
Medchemexpress LLC HY-10619 10mg , Niraparib CAS:1038915-60-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1061910MG
Encompass_Preferred
HY-10619 10mg Niraparib CAS: 1038915-60-4 Niraparib (MK-4827) is a highly potent and orally bioavailable PARP1 and PARP2 inhibitor with IC50s of 3.8 and 2.1 nM, respectively. Niraparib leads to inhibition of repair of DNA damage, activates apoptosis and shows anti-tumor activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-8990