Learn More
Medchemexpress LLC Niraparib tosylate | 1038915-73-9 | MFCD28167748 | 100.0% | 492.6 g/mol | C26H28N4O4S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10619B10MG
Encompass_Preferred
Niraparib tosylate is the tosylate salt of niraparib, a potent, orally bioavailable inhibitor of poly(ADP-ribose) polymerases PARP1 and PARP2, used as a research reagent to study DNA damage repair, apoptosis, and antitumor activity.
  • Potent PARP-2 and PARP-1 inhibition (IC50 2.1 nM and 3.8 nM).
  • High reported purity suitable for analytical and preclinical work (manufacturer reported 99.99%).
  • Tosylate salt form for improved handling and stability.
  • Useful for studies of DNA damage response and apoptosis mechanisms.
  • Available in research-scale quantities for laboratory use.
Catalog No. 50-002-43521