Learn More
Medchemexpress LLC Niraparib tosylate | 1038915-73-9 | MFCD28167748 | 492.59 g·mol⁻¹ | C26H28N4O4S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY10619B100MG
Encompass_Preferred
Niraparib tosylate is the tosylate salt of niraparib, a potent PARP1 and PARP2 inhibitor used in oncology research to inhibit DNA damage repair and induce apoptosis. It is provided for use in in vitro and in vivo studies with documented solubility and storage guidance.
  • Potent PARP1 and PARP2 inhibition (IC50 3.8 nM and 2.1 nM).
  • Suitable for in vitro and in vivo formulations with high DMSO solubility.
  • Tosylate salt form improves handling and solubility.
  • Clear storage recommendations for solid and solution states.
  • Used to study DNA damage response, cell cycle, and apoptosis.
Catalog No. 50-002-43130