Learn More
TARGETMOL CHEMICALS INC NIRAPARIB TOSYLATE 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T689225MG
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Niraparib tosylate (MK-4827 (tosylate))(with IC50 of 3.8 nM/2.1 nM) is a selective PARP1/PARP2 inhibitor. purity: 99%
Catalog No. 50-257-6422