Learn More
Selleck Chemical LLC Niraparib tosylate
SDP
Supplier:  Selleck Chemical LLC S76251G
Encompass_Preferred
Niraparib tosylate is a selective inhibitor of PARP1/PARP2 with IC50 of 3 8 nM/2 1 nM Niraparib increases formation of PARP-DNA complexes resulting in DNA damage apoptosis and cell death
Catalog No. 50-000-00241