Learn More
Medchemexpress LLC Niraparib (tosylate hydrate) | 1613220-15-7 | MFCD28167748 | 100.0% | 510.61 | C26H30N4O5S | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10619E10MG
Encompass_Preferred
Niraparib (tosylate hydrate) is the tosylate hydrate salt of niraparib, a potent PARP1 and PARP2 inhibitor used in research to study DNA damage response and cancer biology. It promotes accumulation of DNA damage and apoptosis in tumor cells and is supplied in formats suitable for in vitro studies.
Catalog No. 50-002-42919