Learn More
Medchemexpress LLC Niraparib tosylate hydrate | 1613220-15-7 | MFCD28167748 | 100.0% | 510.61 g/mol | C26H30N4O5S | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10619E10MM 1ML
Encompass_Preferred
Niraparib tosylate hydrate is the tosylate salt form of niraparib, a potent PARP inhibitor used in biochemical and cell-based research to probe DNA damage response, PARP inhibition, and related combination therapies.
  • Potent PARP1 and PARP2 inhibition (IC50s ~3.8 and 2.1 nM).
  • Supplied as a 10 mM solution in DMSO (1 mL) and as powder formats for flexible dosing.
  • High purity suitable for in vitro studies (product listed as 99.97%).
  • CAS registry number 1613220-15-7 for unambiguous identification.
  • Suitable for cell treatment and biochemical assays investigating DNA repair pathways.
Catalog No. 50-002-42698