Learn More
Medchemexpress LLC Niridazole | 61-57-4 | 98.8% | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1196145MG
Encompass_Preferred
Niridazole is a nitrothiazole-class antibacterial research compound active against anaerobic and microaerophilic bacteria. Supplied as a research-grade solid for in vitro pharmacology, mechanism-of-action studies, and microbiology assays.
  • Antibacterial activity against anaerobic bacteria.
  • Reported IC50 values in the low μg/mL to sub-μg/mL range.
  • High purity suitable for research applications.
  • Soluble in DMSO for assay preparation.
  • Supplied as a small quantity pack for laboratory use.
Catalog No. 50-002-33046