Learn More
Medchemexpress LLC HY-138687 5mg , Nirmatrelvir CAS:2628280-40-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1386875MG
Encompass_Preferred
HY-138687 5mg Nirmatrelvir CAS: 2628280-40-8 Nirmatrelvir (PF-07321332) is a potent and orally active SARS-CoV 3C-like protease (3CLPRO) inhibitor. Nirmatrelvir (PF-07321332) targets to the SARS-CoV-2 virus and can be used for COVID-19 research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-8961