Learn More
Medchemexpress LLC Nirmatrelvir-d9 (PF-07321332-d9) | 2861202-76-6 | 99.9% | 508.58 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY138687S1MG
Encompass_Preferred
Nirmatrelvir-d9 (PF-07321332-d9) is the deuterium labeled Nirmatrelvir. It is intended for research use only and not for sale to patients. This compound can be used as a tracer or as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention due to its potential to impact the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes into drug molecules for quantitation during drug development.
  • Potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Inhibits SARS-CoV-2 3CLPRO, which is responsible for cleaving viral polyproteins and inhibiting viral replication.
Catalog No. 50-003-51244