Learn More
Medchemexpress LLC Nirogacestat | 1290543-63-3 | C27H41F2N5O | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1518510MM/1ML
Encompass_Preferred
Nirogacestat (PF-3084014) is a reversible, orally bioavailable, noncompetitive, and selective γ-secretase inhibitor with an IC50 of 6.2 nM. It inhibits Notch signaling, which is a promising approach for research of Notch receptor-dependent cancers while minimizing gastrointestinal toxicity.
  • Reversible, orally bioavailable, noncompetitive, and selective γ-secretase inhibitor
  • Inhibits Notch signaling
  • Minimizes gastrointestinal toxicity
  • Potential for research of Notch receptor-dependent cancers
Catalog No. 50-003-74144