Learn More
Medchemexpress LLC Nirogacestat | 1290543-63-3 | C27H41F2N5O | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1518550MG
Encompass_Preferred
Nirogacestat is a reversible, orally bioavailable, noncompetitive, and selective γ-secretase inhibitor. With an IC50 of 6.2 nM, it inhibits Notch signaling, minimizing gastrointestinal toxicity, and presents a promising approach for research of Notch receptor-dependent cancers.
  • Reversible, orally bioavailable, noncompetitive, and selective γ-secretase inhibitor.
  • IC50 of 6.2 nM.
  • Inhibits Notch signaling.
  • Minimizes gastrointestinal toxicity.
  • Promising approach for research of Notch receptor-dependent cancers.
Catalog No. 50-003-73258