Learn More
Medchemexpress LLC Nisin | 1414-45-5 | 3354.07 | 1 G
SDP
Supplier:  Medchemexpress LLC HYP16071G
Encompass_Preferred
Nisin is a bacteriocin produced by a group of Gram-positive bacteria, including Lactococcus and Streptococcus species. It exhibits both antibacterial and anti-inflammatory activities, making it suitable for various research applications.
  • Inhibits HNSCC cell ballooning
  • Induces free-living amoeba acanthamoeba cell cycle arrest
  • Reduces uterine inflammation in rats
  • Improved levels of B7-2, IFN-γ, IL-2 and IL-8 in rat models
Catalog No. 50-004-23746