Learn More
Medchemexpress LLC Nisin | 1414-45-5 | 3354.07 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYP1607100MG
Encompass_Preferred
Nisin is a bacteriocin produced by Gram-positive bacteria, specifically from Lactococcus and Streptococcus species. It exhibits both antibacterial and anti-inflammatory activities, making it suitable for various biological applications.
  • Inhibits HNSCC cell ballooning
  • Induces cell cycle arrest in free-living amoeba
  • Reduces uterine inflammation in rats
  • Water soluble
Catalog No. 50-004-23843