Learn More
Medchemexpress LLC Nisin | 1414-45-5 | 3354.07 | 5 G
SDP
Supplier:  Medchemexpress LLC HYP16075G
Encompass_Preferred
Nisin is a bacteriocin produced by Gram-positive bacteria, including Lactococcus and Streptococcus species, and exhibits both antibacterial and anti-inflammatory activities. It is suitable for various research applications.
  • Inhibits HNSCC cell ballooning
  • Induces free-living amoeba cell cycle arrest
  • Reduces uterine inflammation in rats by modulating pro-inflammatory and anti-inflammatory cytokines
Catalog No. 50-004-24015