Learn More
TOKU-E NISIN SOLN 5X1ML
Supplier:  TOKU-E N0845X1ML
Encompass_Preferred
Antibiotic, ReadyMade™, sterile-filtered, 20,000-40,000 IU/ml in 0.02N HCl, Useful in COVID-19 research, cell culture, food safety
Catalog No. NC2068749