Learn More
Apexbio Technology LLC Nisoldipine, 500mg. Cas: 63675-72-9 MFCD: MFCD00478055. calcium channel blocker
SDP
Supplier:  Apexbio Technology LLC B1989500
Encompass_Preferred
Nisoldipine is a calcium channel blocker belonging to the dihydropyridines class, specific for L-type Cav1.2 with IC50 of 10 nM. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5296