Learn More
Medchemexpress LLC Nisoxetine (hydrochloride) | 57754-86-6 | 99.92% | 10 MM * 1 ML
SDP
Supplier:  Medchemexpress LLC HYB1704A10MM/1ML
Encompass_Preferred
Nisoxetine hydrochloride is a potent and selective inhibitor of noradrenaline transporter (NET) with a Kd of 0.76 nM. It is an antidepressant and local anesthetic that can block voltage-gated sodium channels.
  • Potent and selective inhibitor of noradrenaline transporter (NET)
  • Kd of 0.76 nM (NET)
  • Antidepressant
  • Local anesthetic
  • Blocks voltage-gated sodium channels
Catalog No. 50-004-13175