Learn More
Cayman Chemical NIsoxetInhydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 2963710 MG
Encompass_Preferred
A NET inhibitor (Ki = 5.1 nM); selective for NET over DAT and SERT (Kis = 477 and 383 nM, respectively); inhibits norepinephrine uptake and inhibits amphetamine-induced increases in norepinephrine release in mouse brain (EC50s = 2.97 and 0.08 µM, respectively); reduces increases in locomotor activity induced by d-N-ethylamphetamine, methylphenidate, and cocaine, but not morphine, in mice at 32 mg/kg; reduces cataplexy in narcoleptic dogs at 0.5 mg/kg i.v.
Catalog No. 50-298-8898