Learn More
Cole-Parmer Fowler 52-262-624 NIST Calibration Certificate Rigid-Ruler Set, 6", 12" and 24" Rules
Supplier:  Cole-Parmer 0991932
Encompass_Preferred
Certified rulers are made of hardened and tempered steel and feature easy to read graduations on a non-glare satin chrome finish. Each Fowler Ruler is Pre-Certified for ISO/QS9000 requirements and includes a certificate of calibration.
Catalog No. NC0885239