Learn More
Thermco Products Inc NIST Trcble N-Mercury Thermometer
Supplier: Thermco Products Inc ACC1005SFC
Thermco Certified Thermometers are used in temperature verification and calibration procedures. These instruments can be used to calibrate Liquid-In- Glass, RTD, Thermistors, Thermocouples, and Bi-Metal Thermometers. The certified thermometers are carefully inspected to insure they conform to exact specifications and are certified against NlST standards. Each thermometer is furnished with a document proving traceability and a felt-lined leatherette storage case.