Learn More
Selleck Chemical LLC Nitazoxanide 100mg 55981-09-4
SDP
Supplier:  Selleck Chemical LLC S1627100MG
Encompass_Preferred
Nitazoxanide is a synthetic nitrothiazolyl-salicylamide derivative and an antiprotozoal agent(IC50 for canine influenza virus ranges from 0.17 to 0.21 ?M). Nitazoxanide modulates autophagy and inhibits mTORC1 signaling. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-539-0