Learn More
Cayman Chemical NItazoxnIde 10mg
SDP
Supplier:  Cayman Chemical 1369210 MG
Encompass_Preferred
A prodrug form of tizoxanide; inhibits C. parvum growth in MDBK cells at 10 UG/ml; active against M. tuberculosis and M. bovis (MIC = 16 UG/ml for both); inhibits SARS-CoV-2 and MERS-CoV replication at 5 and 10 µM; inhibits replication of H1N1 PR8 and WSN, as well as H5N9 A/Ck viruses (EC50s = 1, 0.5, and 1 UG/ml, respectively); reduces oocyst shedding and parasite burden, but not the severity of diarrhea, in a gnotobiotic piglet model of C. parvum infection at 250 mg/kg
Catalog No. 50-295-3452