Learn More
Medchemexpress LLC NITD-688 | 2407227-31-8 | 98.04% | 500.68 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY13854310MM/1ML
Encompass_Preferred
NITD-688, also known as EYU-688, is an orally active pan-serotype inhibitor of the dengue virus NS4B protein. It is suitable for research related to the dengue virus (DENV).
  • Shows antiviral activity against all four serotypes of dengue virus (DENV)
  • Directly binds to wild-type NS4B protein
  • Demonstrates excellent antiviral efficacy in infected AG129 mice
  • Well tolerated in rats and dogs in toxicology studies
Catalog No. 50-003-48389