Learn More
Medchemexpress LLC NITD008 | 1044589-82-3 | 98.00% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY12957
Encompass_Preferred
NITD008 is a potent and selective flavivirus inhibitor that can inhibit Dengue Virus Type 2 (DENV-2) with an EC50 of 0.64 μM. It is also a click chemistry reagent containing an Alkyne group, allowing it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that have Azide groups.
  • Potent and selective flavivirus inhibitor
  • Inhibits Dengue Virus Type 2 (DENV-2) with an EC50 of 0.64 μM
  • Click chemistry reagent with Alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
Catalog No. 50-003-42688