Learn More
ELKO FILTERING CO NITEX SHEETS 110UM 1 YARD
SDP
Supplier:  ELKO FILTERING CO 03110/47
Encompass
Nylon Monofilament Precision Woven Mesh - 110 Micron Opening, Open Area 47%, 40 Inch wide, Sold by Yard - Available from 1 - 5000 Microns - Sefar 03-110/47
Catalog No. NC0123567