Learn More
ELKO FILTERING CO NITEX SHEETS 50UM 1 YARD
SDP
Supplier:  ELKO FILTERING CO 0350/37
Encompass
Nylon Monofilament Precision Woven Mesh - 50 Micron Opening, Open Area 37%, 40 Inch wide, Sold by Yard - Available from 1 - 5000 Microns - Sefar 03-50/37
Catalog No. NC0486649