Learn More
Medchemexpress LLC Nitisinone | 104206-65-7 | 99.93% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB06075MG
Encompass_Preferred
Nitisinone is an orally active, competitive, and reversible 4-hydroxyphenylpyruvate dioxygenase (4-HPPD) inhibitor. It promotes tyrosine accumulation in a dose-dependent manner and is utilized in studies of hereditary tyrosinemia type 1 (HT-1) and albinism.
  • Orally active 4-HPPD inhibitor
  • Promotes tyrosine accumulation
  • Used in studies of hereditary tyrosinemia type 1 (HT-1)
  • Applicable in albinism research
  • Purity of 99.93%
Catalog No. 50-004-07775