Learn More
Selleck Chemical LLC Nitisinone (NTBC) S5325-1g
SDP
Supplier:  Selleck Chemical LLC S53251G
Encompass_Preferred
Nitisinone (NTBC) is a synthetic reversible inhibitor of 4-hydroxyphenylpyruvate dioxygenase used in the treatment of hereditary tyrosinemia type 1
Catalog No. 50-311-2304