Learn More
Cayman Chemical Nitrate/Nitrite Fluorometric Assay Kit, 2 x 96 wells
SDP
Supplier:  Cayman Chemical 7800512X96WELLS
Encompass_Preferred
Cayman’s Nitrate/Nitrite Fluorometric Assay provides a convenient method for the quantitation of low levels of nitrate and nitrite in biological samples (particularly tissue culture medium). Previously undetectable concentrations of nitrate/nitrite in a variety of samples can be accurately quantitated using this kit as it extends the lower limit of detection by 20-fold over our colorimetric version (Item No. 780001). The minimum detectable quantity of NO2/NO3 is ~50 nM.
Catalog No. NC9921093
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.