Learn More
GFS Chemicals Inc NITRIC ACID, 70%, ENVIRONMENTAL GRADE | 6 X 500 ML | GFS CHEMICALS, INC. Item #: 53080 | Bulk #: 2543
SDP
Supplier:  GFS Chemicals Inc 53080
Encompass_Preferred
NITRIC ACID, 70%| Specs Max : Ag 1.0 ppb , Al 1.0 ppb , As 1.0 ppb , Assay 68.0 - 70.0 %, Au 1.0 ppb , B 5.0 ppb , Ba 1.0 ppb , Be 1.0 ppb , Bi 1.0 ppb , Ca 5.0 ppb , Cd 1.0 ppb , Ce 1.0 ppb , Chloride 0.00001 %, Co 1.0 ppb , Color (APHA) 10 , Cr 1.0 ppb , Cs 1.0 ppb , Cu 1.0 ppb , Dy 1.0 ppb , Er 1.0 ppb , Eu 1.0 ppb , Ga 1.0 ppb , Gd 1.0 ppb , Ge 1.0 ppb , Hf 1.0 ppb , Hg 2.0 ppb , Ho 1.0 ppb , In 1.0 ppb , Ir 1 ppb , Iron 0.0000001 % , K 1.0 ppb , La 1.0 ppb , Li 1.0 ppb , Lu 1.0 ppb , Mg 1.0 ppb , Mn 1.0 ppb , Mo 1.0 ppb , Na 3.0 ppb , Nd 1.0 ppb , Ni 1.0 ppb , Pb 1.0 ppb , Pd 1.0 ppb , Pr 1.0 ppb , Pt 1.0 ppb , Rb 1.0 ppb , Residue after ignition 0.0001 % , Rh 1.0 ppb , Ru 1.0 ppb , Sb 1.0 ppb , Sc 1.0 ppb , Se 1.0 ppb , Sm 1.0 ppb , Sn 1.0 ppb , Sr 1.0 ppb , Sulfate 0.00001 % , Tb 1.0 ppb , Te 1.0 ppb , Th 1.0 ppb , Ti 1.0 ppb , U 1.0 ppb , V 1.0 ppb , W 1.0 ppb , Y 1.0 ppb , Yb 1.0 ppb , Zn 1.0 ppb , Zr 1.0 ppb
Catalog No. NC1525008
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.