Learn More
Chemglass Life Sciences Nitrile Gloves, Sky Blue, Extra Small, Examination, Protege
SDP
Supplier:  Chemglass Life Sciences CG1790P01
Encompass_Preferred
Nitrile Gloves, Sky Blue, Extra Small, Examination, Prot¿. The Aurelia Prot¿ nitrile, powder-free, disposable glove is made from stretch nitrile and is textured along the length of the fingers for improved grip compared to a glove with textured fingertips. The glove has a palm thickness of 4 mils and a finger thickness of 5.2 mils, and is 9.5" long from the tip of the middle finger to the edge of the beaded wrist cuff, which is 2 mils thick. Nitrile provides better elasticity than vinyl, better puncture resistance than latex or vinyl, better abrasion resistance than chloroprene, and reduces the likelihood of allergic reaction. This powder-free glove leaves no residue, reducing the risk of contamination compared to a powdered glove. This ambidextrous blue glove is suitable for dental and medical examination applications.
Catalog No. 50-197-9494