Learn More
Medchemexpress LLC Nitrilotriacetic acid | 139-13-9 | MFCD00004287 | 98.0% | 191.14 g/mol | C6H9NO6 | 1 G
SDP
Supplier:  Medchemexpress LLC HYW0307781G
Encompass_Preferred
Nitrilotriacetic acid (CAS 139-13-9) is an aminotricarboxylic acid used as a chelating agent and analytical reference standard. It is supplied as a solid for laboratory and analytical applications and is suitable for standard preparation and research use. Molecular formula C6H9NO6; molecular weight 191.14 g/mol.
  • High-purity analytical standard (98.0%).
  • Effective chelating agent for metal ion sequestration.
  • Supplied as a solid suitable for formulation or standard preparation.
  • Available in small laboratory pack sizes for analytical use.
  • Molecular formula C6H9NO6 and molecular weight 191.14 g/mol.
Catalog No. 50-003-00193