Learn More
TARGETMOL CHEMICALS INC NITROBENZYLTHIOINOSINE 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T855125MG
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Nitrobenzylthioinosine (NBMPR) is an inhibitor of ENT1 transporter that binds to ENT1 transporter with high affinity. purity: 99%
Catalog No. 50-257-6235