Learn More
Selleck Chemical LLC Nitrocaramiphen hydrochloride
SDP
Supplier:  Selleck Chemical LLC E08295MG
Encompass_Preferred
Nitrocaramiphen hydrochloride is an antagonist of muscarinic acetylcholine receptor M1 (mAChR M1) which can completely remove the hyperpolarizing effect of muscarine
Catalog No. 50-313-8817