Learn More
Apexbio Technology LLC Nitrocefin 41906-86-9 25mg
SDP
Supplier:  Apexbio Technology LLC B605225
Encompass_Preferred
Nitrocefin is a chromogenic cephalosporin substrate commonly employed in biochemical assays to detect -lactamase enzymatic activity -lactamases enzymes prevalent in microbial species catalyze the hydrolysis of -lactam antibiotics such as penicillins and cephalosporins causing antibiotic resistance Upon cleavage by -lactamase enzymes nitrocefin undergoes a measurable colorimetric shift from yellow to red enabling visual or spectrophotometric detection in the range of approximately 380 500 nm Nitrocefin prominently serves in microbiological and clinical research to evaluate resistance profiles of bacterial isolates and in assays for inhibitor activity toward -lactamases Its reported IC50 varies depending on -lactamase type concentration and assay conditions with typical values ranging from approximately 0 5 to 25 M
Catalog No. 50-000-63647