Learn More
Medchemexpress LLC HY-A0090 500mg Medchemexpress, Nitrofurantoin CAS:67-20-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYA0090500MG
Encompass_Preferred
Medchemexpress, HY-A0090 500mg Nitrofurantoin CAS:67-20-9 Nitrofurantoin is a potent and orally active broad-spectrum beta-lactamase antimicrobial agent. Nitrofurantoin acts as an antibiotic and can be used for the study of urinary tract infections (UTIs), including cystitis and kidney infections. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2324