Learn More
Medchemexpress LLC Nitrofurazone | 1217220-85-3 | 99.6% | 201.12 g/mol | (13C)C5H6(15N)2N2O4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB0226S10MG
Encompass_Preferred
Nitrofurazone-13C,15N2 is a stable isotope-labeled form of the antibacterial nitrofurazone, supplied as a high-purity analytical standard for use in mass spectrometry, tracer studies, and quantitative assays. The labeled product contains one 13C and two 15N atoms and has a molecular weight of 201.12 g/mol and a reported chemical purity of 99.56%.
  • Stable isotope labeled for use as an internal standard and tracer.
  • High chemical purity suitable for analytical workflows.
  • Available in small, weighed quantities for laboratory use.
  • Provided as a solid with defined molecular weight and CAS number for traceability.
Catalog No. 50-002-98991