Learn More
Chemglass Life Sciences Chemglass Nitrogen Flowmeter CG-1605-A-12, 0.4 to 5.0 L/min
SDP
Supplier:  Chemglass Life Sciences CG1605A12
Encompass_Preferred
Precision acrylic nitrogen flowmeter with an integrated brass metering valve designed to meter nitrogen gas for laboratory process reactors and low-flow gas lines. Provides adjustable control across a low-flow range suitable for small-scale reactions and inert gas blanketing.
  • Precision clear acrylic body for visual flow indication.
  • Brass metering valve for fine flow control.
  • Buna-N seals compatible with common laboratory gases.
  • Flow range 0.4 to 5.0 L/min for low-flow applications.
  • Designed for use with process reactors and gas delivery lines.
  • Supplied individually (each).
Catalog No. NC2076263