Learn More
ASI STANDARDS NITROGEN SOLID POLY 15000MG/KG
Supplier:  ASI STANDARDS NP1500025
Encompass_Preferred
Nitrogen in Solid Polymer for Quality Control, Concentration 15000 mg/kg, 25 g
Catalog No. NC1943001