Learn More
Hach Company Nitrogen (Total) TNTplus Vial Test, LR (1-16 mg/L N), 25 Tests
Supplier:  Hach Company TNT826
Encompass
Nitrogen TNTplus chemistry and your Hach spectrophotometer are engineered to simplify water analysis for accurate results, everytime.Hach Method 10208 determines Total Nitrogen in water and wastewater by the Persulfate Digestion Method with a range up to 16 mg/L N. Digestion occurs within the sealed vial which eliminates evaporation and nitrogen loss due to splatter.
Catalog No. 50-199-8122